Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT4, Mouse, Clone: AHN1.1, Abnova™

Catalog No. 89113070 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-070 Supplier Abnova Corporation Supplier No. MAB6931
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native FUT4.

The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). [provided by RefSeq

Specifications

Antigen FUT4
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone AHN1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native FUT4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate)
Gene FUT4
Gene Alias CD15/ELFT/FCT3A/FUC-TIV/FUTIV
Gene Symbols FUT4
Host Species Mouse
Immunogen Native purified FUT4 from human neutrophils.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2526
Test Specificity Antibody AHN1.1 activates normal monocytes and inhibits neutrophil chemotaxis.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.