Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fut4, Mouse, Clone: MC-480, Abnova™

Catalog No. 89110995 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-995 Supplier Abnova Corporation Supplier No. MAB2413
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Fut4.

Specifications

Antigen Fut4
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MC-480
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Fut4.
Dilution Flow Cytometry (1:50-1:200) Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene Fut4
Gene Alias AI451562/FAL/FucT-IV/LeX
Gene Symbols Fut4
Host Species Mouse
Immunogen Native purified from F9 tetracarcinoma stem cells (X-irradiated).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14345
Target Species Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.