Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fxyd1, Mouse, Clone: 41AT858.235, Abnova™

Catalog No. 89107882 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-882 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-882 Supplier Abnova Corporation Supplier No. MAB0116
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of Fxyd1.

Specifications

Antigen Fxyd1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 41AT858.235
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of Fxyd1.
Dilution Western Blot (1:100-500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Fxyd1
Gene Alias 0610012C17Rik/1110006M24Rik/PLM/PML
Gene Symbols Fxyd1
Host Species Mouse
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S68 of mouse Fxyd1.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56188
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.