Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fyn, Mouse, Clone: 1S, Abnova™

Catalog No. 89110687 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-687 Supplier Abnova Corporation Supplier No. MAB1882

Mouse monoclonal antibody raised against partial recombinant Fyn.

Specifications

Antigen Fyn
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1S
Conjugate Unconjugated
Description Fyn proto-oncogene
Formulation In PBS (0.08% sodium azide)
Gene Fyn
Gene Alias AI448320, AW552119, MGC115870
Gene Symbols Fyn
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-206 of mouse Fyn.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14360
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.