Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD4, Mouse, Clone: 3G7, Abnova™

Catalog No. 89105834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-834 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-834 Supplier Abnova Corporation Supplier No. H00008322M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FZD4.

This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. [provided by RefSeq

Sequence: TEKINIPIGPCGGMCLSVKRRCEPVLKEFGFAWPESLNCSKFPPQNDHNHMCMEGPGDEEVPLPHKTPIQPGEECHSVGTNSDQYIWVKRSLNCVLKCGY

Specifications

Antigen FZD4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FZD4.
Formulation ascites with no preservative
Gene FZD4
Gene Accession No. NM_012193
Gene Alias CD344/EVR1/FEVR/FZD4S/Fz-4/FzE4/GPCR/MGC34390
Gene Symbols FZD4
Host Species Mouse
Immunogen FZD4 (NP_036325,107 a.a. ∽ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8322
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.