Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-162 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-162 Supplier Abnova Corporation Supplier No. H00009846B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GAB2 protein.

This gene is a member of the GRB2-associated binding protein (GAB) gene family and is similar to the GAB1 gene. These proteins contain pleckstrin homology (PH) domain, and bind SHP2 tyrosine phosphatase and GRB2 adapter protein. They act as adapters for transmitting various signals in response to stimuli through cytokine and growth factor receptors, and T- and B-cell antigen receptors. The protein encoded by this gene is the principal activator of phosphatidylinositol-3 kinase in response to activation of the high affinity IgE receptor. Two alternatively spliced transcripts encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: MSGGGDVVCTGWLRKSPPEKKLRRYAWKKRWFILRSGRMSGDPDVLEYYKNDHSKKPLRIINLNFCEQVDAGLTFNKKELQDSFVFDIKTSERTFYLVAETEEDMNKWVQSICQICGFNQAEESTDSLRNVSSAGHGPRSSPAELSSSSQHLLRERKSSAPSHSSQPTLFTFEPPVSNHMQPTLSTSAPQEYLYLHQCISRRAENARSASFSQGTRASFLMRSDTAVQKLAQGNGHCVNGISGQVHGFYSLPKPSRHNTEFRDSTYDLPRSLASHGHTKGSLTGSETDNEDVYTFKTPSNTLCREFGDLLVDNMDVPATPLSAYQIPRTFTLDKNHNAMTVATPGDSAIAPPPRPPKPSQAETPRWGSPQQRPPISENSRSVAATIPRRNTLPAMDNSRLHRASSCETYEYPQRGGESAGRSAESMSDGVGSFLPGKMIVGRSDSTNSEDNYVPMNPGSSTLLAMERAGDNSQSVYIPMSPGAHHFDSLGYPSTTLPVHRGPSRGSEIQPPPVNRNLKPDRKAKPTPLDLRNNTVIDELPFKSPITKSWSRANHTFNSSSSQYCRPISTQSITSTDSGDSEENYVPMQNPVSASPVPSGTNSPAPKKSTGSVDYLALDFQPSSPSPHRKPSTSSVTSDEKVDYVQVDKEKTQALQNTMQEWTDVRQSSEPSKGAKL

Specifications

Antigen GAB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GAB2 protein.
Formulation PBS with no preservative; pH 7.4
Gene GAB2
Gene Accession No. NM_080491
Gene Alias KIAA0571
Gene Symbols GAB2
Host Species Mouse
Immunogen GAB2 (NP_536739,1 a.a. ∽ 676 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9846
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.