Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABPB1, Mouse, Clone: 10, Abnova™

Catalog No. 89113877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-877 Supplier Abnova Corporation Supplier No. MAB9919
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant GABPB1.

This gene encodes the GA-binding protein transcription factor, beta subunit. This protein forms a tetrameric complex with the alpha subunit, and stimulates transcription of target genes. The encoded protein may be involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. The crystal structure of a similar protein in mouse has been resolved as a ternary protein complex. Multiple transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Specifications

Antigen GABPB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant GABPB1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene GABPB1
Gene Accession No. NM_016655
Gene Alias BABPB2/E4TF1/E4TF1-47/E4TF1-53/E4TF1B/GABPB/GABPB2/NRF2B1/NRF2B2
Gene Symbols GABPB1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human GABPB1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2553
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.