Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabra1, Mouse, Clone: S95-35, Abnova™

Catalog No. 89113227 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-227 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-227 Supplier Abnova Corporation Supplier No. MAB7906
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Gabra1.

Specifications

Antigen Gabra1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S95-35
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Gabra1.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Gabra1
Gene Alias Gabra-1
Gene Symbols Gabra1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 355-394 of mouse Gabra1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14394
Test Specificity No cross-reactivity against GABA-A-R-Alpha2 or -Alpha3.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.