Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNT10, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-043 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-043 Supplier Abnova Corporation Supplier No. H00055568A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GALNT10.

This gene belongs to the polypeptide N-acetylgalactosaminyltransferase (pp-GalNAc-T) gene family. Polypeptide GalNAc transferases initiate the synthesis of mucin-type oligosaccharides by transferring GalNAc from UDP-GalNAc to the hydroxyl group of either a serine or threonine residue on the polypeptide acceptor. Following expression in insect cells, recombinant GalNAc transferase 10 showed significant GalNAcT activity toward mucin-derived peptides, and it utilized both nonglycosylated and glycosylated peptide substrates. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: FTFTWREDIRPGDPQHTKKFCFDAISHTSPVTLYDCHSMKGNQLWKYRKDKTLYHPVSGSCMDCSESDHRIFMNTCNPSSLTQQWLFEHTNSTVLEKFNR

Specifications

Antigen GALNT10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GALNT10.
Formulation 50% glycerol
Gene GALNT10
Gene Accession No. NM_198321
Gene Alias DKFZp586H0623/FLJ00205/FLJ11715/GalNAcT10/pp-GalNAc-T10
Gene Symbols GALNT10
Host Species Mouse
Immunogen GALNT10 (NP_938080, 503 a.a. to 602 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55568
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.