Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNT7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106937 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-937 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-937 Supplier Abnova Corporation Supplier No. H00051809A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GALNT7.

This gene encodes GalNAc transferase 7, a member of the GalNAc-transferase family. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. Unlike other family members, this enzyme shows exclusive specificity for partially GalNAc-glycosylated acceptor substrates and shows no activity with nonglycosylated peptides. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation. [provided by RefSeq

Sequence: LGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSV

Specifications

Antigen GALNT7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GALNT7.
Formulation 50% glycerol
Gene GALNT7
Gene Accession No. NM_017423
Gene Alias GALNAC-T7/GalNAcT7
Gene Symbols GALNT7
Host Species Mouse
Immunogen GALNT7 (NP_059119, 559 a.a. to 657 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51809
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.