Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNTL4, Mouse, Clone: 1F9, Abnova™

Catalog No. 89107779 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-779 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-779 Supplier Abnova Corporation Supplier No. H00374378M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GALNTL4.

Sequence: SQQIHVGILSPTVDDDDNRCLVDVNSRPRLTECSYAKAKRMKLHWQFSQGGPIQNRKSKRCLELQENSDLEFGFQLVLQKCSGQHWSITNVLRSL*

Specifications

Antigen GALNTL4
Applications ELISA
Classification Monoclonal
Clone 1F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GALNTL4.
Formulation ascites with no preservative
Gene GALNTL4
Gene Accession No. NM_198516
Gene Alias GALNT15/GALNT18/GalNAc-T15/MGC71806
Gene Symbols GALNTL4
Host Species Mouse
Immunogen GALNTL4 (NP_940918,511 a.a. ∽ 606 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374378
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.