Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-gamma-Actin/ACTG1, Clone: 2A3,

Catalog No. MABT824 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT824 Supplier MilliporeSigma™ Supplier No. MABT824
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects gamma-Actin/ACTG1 clone: 2A3 in Chicken, Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Western Blotting

Actin, cytoplasmic 2 (UniProt P63261; also known as Gamma-actin) is encoded by the ACTG1 (also known as ACTG, BRWS2, DFNA20, DFNA26) gene (Gene ID 71) in human. Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin is encoded by the ACTA1 gene, smooth muscle alpha-actin by the ACTA2 gene, cytoplasmic beta-actin by the ACTB gene, cardiac muscle alpha-actin by the ACTC1 gene, cytoplasmic gamma-actin by the ACTG1 gene, and smooth muscle gamma-actin by the ACTG2 (a.k.a. ACTA3) gene. Although actins show 90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic β and γ-actins, are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma-Actin/ACTG1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2A3
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P63261
Gene Symbols ACTG1; ACTG; BRWS2; DFNA20; DFNA26
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the the N-terminal sequence of human gamma-Actin/ACTG1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001605.1
Target Species Chicken, Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.