Learn More
Specifications
Specifications
| Antigen | GAP43 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1G7 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against full length recombinant GAP43. |
| Dilution | Immunohistochemistry (1:20-1:40) The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from tissue culture supernatant (0.09% sodium azide) |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to full length GAP43. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.