Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH, Mouse, Clone: 10B8, Abnova™

Catalog No. 89111134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-134 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-134 Supplier Abnova Corporation Supplier No. MAB2781

Mouse monoclonal antibody raised against GAPDH.

Specifications

Antigen GAPDH
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 10B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against GAPDH.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias G3PD/GAPD/MGC88685
Gene Symbols GAPDH
Host Species Mouse
Immunogen Human or rabbit GAPDH.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2597/100009074
Target Species Human, Rabbit
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.