Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH, Mouse, Clone: 1D4, Abnova™

Catalog No. 89110928 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-928 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-928 Supplier Abnova Corporation Supplier No. MAB2319

Mouse monoclonal antibody raised against GAPDH.

Specifications

Antigen GAPDH
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against GAPDH.
Dilution Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (10mM sodium azide)
Gene GAPDH
Gene Alias GAPD
Gene Symbols GAPDH
Host Species Mouse
Immunogen Pig GAPDH.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396823
Test Specificity On Western Blot reveals a prominent ∼37 KDa band.
Target Species Avian, Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.