Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH, Mouse, Clone: AT8G4, Abnova™

Catalog No. 89113522 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-522 Supplier Abnova Corporation Supplier No. MAB8701
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant GAPDH.

The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. [provided by RefSeq

Specifications

Antigen GAPDH
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AT8G4
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant GAPDH.
Dilution Western Blot (1:500-1:10000) Immunofluorescence (1:500-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene GAPDH
Gene Alias G3PD/GAPD/MGC88685
Gene Symbols GAPDH
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human GAPDH.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2597
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.