Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS7, Mouse, Clone: AT4H8, Abnova™

Catalog No. 89112235 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-235 Supplier Abnova Corporation Supplier No. MAB5376
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant GAS7.

Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described. [provided by RefSeq

Specifications

Antigen GAS7
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT4H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant GAS7.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene GAS7
Gene Alias KIAA0394/MGC1348/MLL/GAS7
Gene Symbols GAS7
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human GAS7.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8522
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.