Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA2, Mouse, Clone: 2F12, Abnova™

Catalog No. 89104535 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-535 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-535 Supplier Abnova Corporation Supplier No. H00002624M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GATA2.

This gene encodes a member of the GATA family of zinc-finger transcription factors that are named for the consensus nucleotide sequence they bind in the promoter regions of target genes. The encoded protein plays an essential role in regulating transcription of genes involved in the development and proliferation of hematopoietic and endocrine cell lineages. Alternative splicing results in multiple transcript variants

Sequence: MEVAPEQPRWMAHPAVLNAQHPDSHHPGLAHNYMEPAQLLPPDEVDVFFNHLDSQGNPYYANPAHARARVSYSPAHARLTGGQMCRPHLLHSPGLPWLDGGK

Specifications

Antigen GATA2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GATA2.
Formulation ascites with no preservative
Gene GATA2
Gene Accession No. BC018988
Gene Alias MGC2306/NFE1B
Gene Symbols GATA2
Host Species Mouse
Immunogen GATA2 (AAH18988,1 a.a. ∽ 102 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2624
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.