Learn More
GCG, Mouse, Clone: 2F9, Abnova™
Mouse monoclonal antibody raised against partial recombinant GCG.
Supplier: Abnova Corporation MAB10665
Description
The protein encoded by this gene is actually a preproprotein that is cleaved into four distinct mature peptides. One of these, glucagon, is a pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis. Glucagon is a ligand for a specific G-protein linked receptor whose signalling pathway controls cell proliferation. Two of the other peptides are secreted from gut endocrine cells and promote nutrient absorption through distinct mechanisms. Finally, the fourth peptide is similar to glicentin, an active enteroglucagon. [provided by RefSeq
Specifications
GCG | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
GCG | |
GCG | |
Recombinant protein corresponding to human GCG. | |
RUO | |
2641 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Western Blot | |
2F9 | |
Mouse monoclonal antibody raised against partial recombinant GCG. | |
In ascites (0.03% sodium azide) | |
GLP1/GLP2/GRPP | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.