Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF15, Mouse, Clone: 23B3D2.H5, Abnova™

Catalog No. 89113366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-366 Supplier Abnova Corporation Supplier No. MAB8161
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of GDF15 .

Bone morphogenetic proteins (e.g., BMP5; MIM 112265) are members of the transforming growth factor-beta (see TGFB1; MIM 190180) superfamily and regulate tissue differentiation and maintenance. They are synthesized as precursor molecules that are processed at a dibasic cleavage site to release C-terminal domains containing a characteristic motif of 7 conserved cysteines in the mature protein.[supplied by OMIM

Specifications

Antigen GDF15
Applications ELISA, Western Blot
Classification Monoclonal
Clone 23B3.D2.H5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of GDF15 .
Dilution ELISA (1:2000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene GDF15
Gene Alias GDF-15/MIC-1/MIC1/NAG-1/PDF/PLAB/PTGFB
Gene Symbols GDF15
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus end of human GDF15 .
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9518
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.