Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP, Mouse, Biotin, Clone: SB61b, Abnova™

Catalog No. 89112530 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-530 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-530 Supplier Abnova Corporation Supplier No. MAB5898
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant GFAP.

This gene encodes one of the major intermediate filament proteins of mature astrocytes. It is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq

Specifications

Antigen GFAP
Applications Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone SB61b
Conjugate Biotin
Description Mouse monoclonal antibody raised against recombinant GFAP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GFAP
Gene Alias FLJ45472
Gene Symbols GFAP
Host Species Mouse
Immunogen Recombinant protein corresponding to human GFAP.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2670
Test Specificity human GFAP (∼ 50 KDa).
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.