Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP, Mouse, Clone: GF-01, Abnova™

Catalog No. 89111500 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-500 Supplier Abnova Corporation Supplier No. MAB3612
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native GFAP.

Specifications

Antigen GFAP
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GF-01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native GFAP.
Dilution Immunocytochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene GFAP
Gene Symbols GFAP
Host Species Mouse
Immunogen Native purified porcine GFAP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396562
Test Specificity This antibody reacts with GFAP, the principal marker of astroglial cells in the central nervous system, which is specifically expressed in satellite cells in peripheral ganglia and in non myelinating Schwann cells in peripheral nerves. The GFAP protein runs on gels at ∼55 KDa protein, usually associated with lower Mw bands which are thought to be proteolytic fragments and alternate transcripts from the single gene.
Target Species Feline, Human, Pig
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.