Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP, Mouse, Clone: MIG-G2, Abnova™

Catalog No. 89135452 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-452 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-452 Supplier Abnova Corporation Supplier No. MAB3540

Mouse monoclonal antibody raised against GFAP.

Specifications

Antigen GFAP
Applications Immunohistochemistry
Classification Monoclonal
Clone MIG-G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against GFAP.
Dilution Immunohistochemistry (1:10-1:25) The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer, pH 7.3 (0.09% sodium azide)
Host Species Mouse
Immunogen Purified Glial Fibrillary Acidic Protein.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody has no cross-reactivity with other filament proteins.
Target Species Guinea Pig, Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.