Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Mouse, Biotin, Clone: 1A12-6-18, BD

Catalog No. BDB565271 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565271 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565271 Supplier BD Biosciences Supplier No. 565271
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody 1A12-6-18 reacts with Green Fluorescent Protein (GFP), which is a 27 kDa bioluminescent protein first purified from the jellyfish, Aequorea victoria . GFP exhibits green fluorescence when exposed to blue or ultraviolet light. Additionally, GFP can be introduced into the genome of and expressed in a wide variety of cell types, making it a useful reporter for tracking gene expression or protein localization. A number of GFP mutants have been developed with varying fluorescence intensity and spectra. Monoclonal antibody 1A12-6-18 is known to react with AcGFP, and is not reactive towards ZsGreen1.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1A12-6-18
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.