Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP tag, Mouse, Clone: 946as, Abnova™

Catalog No. 89136600 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-600 Supplier Abnova Corporation Supplier No. MAB8758
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of GFP tag.

Specifications

Antigen GFP tag
Applications Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 946as
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of GFP tag.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide (conjugated with carrier KLH) corresponding to N-terminus amino acids of Aequorea victoria GFP tag.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes native and denatured forms of GFP and its variants EGFP, YFP, EYFP, and CFP.
Target Species Jellyfish
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.