Learn More
GIMAP5, Mouse, Clone: 3F11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GIMAP5.
Supplier: Abnova Corporation H00055340M14A
Description
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1. [provided by RefSeq
Sequence: MGGFQRGKYGTMAEGRSEDNLSATPPALRIILVGKTGCGKSATGNSILGQPAFESKLRAQSVTRTCQVKTGTWNGRKVLVVDTPSIFESQADTQELYKNIGDCYLLSAPGPHVLLLVIQLGRFTAQDTVAIRKVKEVFGTGAMRHVVILFTHKEDLGGQALDDYVANTDNCSLKDLVRECERRYCAFNNWGSVEEQRQQQAELLAVIERLGREREGSFHSNDLFLDAQLLQRTGAGACQEDYRQYQAKVEWQVEKHKQELRENESNWAYKALLRVKHLMLLHYEIFVFLLLCSILFFIIFLFIFHYISpecifications
GIMAP5 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC011732 | |
GIMAP5 | |
GIMAP5 (AAH11732,1 a.a. ∽ 307 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
55340 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Immunoprecipitation, Western Blot | |
3F11 | |
Mouse monoclonal antibody raised against a full-length recombinant GIMAP5. | |
GIMAP5 | |
FLJ11296/HIMAP3/IAN4/IAN4L1/IAN5/IMAP3/hIAN5 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.