Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC2, Mouse, Clone: AT1G10, Abnova™

Catalog No. 89109358 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-358 Supplier Abnova Corporation Supplier No. MAB11190
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant GIPC2.

Specifications

Antigen GIPC2
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1G10
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant GIPC2.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) with 0.09% sodium azide.
Gene GIPC2
Gene Alias FLJ20075/SEMCAP-2/SEMCAP2
Gene Symbols GIPC2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-315 of human GIPC2.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54810
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.