Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GKN1, Mouse, Clone: 22, Abnova™

Catalog No. 89113881 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-881 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-881 Supplier Abnova Corporation Supplier No. MAB9923
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant GKN1.

The protein encoded by this gene is found to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. [provided by RefSeq

Specifications

Antigen GKN1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 22
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant GKN1.
Dilution Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene GKN1
Gene Accession No. NM_019617.3
Gene Alias AMP18/BRICD1/CA11/FOV/MGC70354/foveolin
Gene Symbols GKN1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 91-199 of human GKN1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56287
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.