Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLO1, Mouse, Clone: Glo1a, Abnova™

Catalog No. 89113143 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-143 Supplier Abnova Corporation Supplier No. MAB7193
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant GLO1.

The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere. [provided by RefSeq

Specifications

Antigen GLO1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone Glo1a
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant GLO1.
Dilution ELISA (50 ug/1 ug protein) Immunofluorescence (10 ug/mL) Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene GLO1
Gene Accession No. Q04760
Gene Alias GLOD1/GLYI
Gene Symbols GLO1
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to full length human GLO1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2739
Test Specificity GLO1 (Glo1a). A band is seen ∼21 KDa in WB.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.