Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Oxidase Mouse anti-Bacteria, Fungi, Monoclonal Antibody, Abnova™

Catalog No. 89136103 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-103 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-103 Supplier Abnova Corporation Supplier No. MAB6146
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Glucose Oxidase.

Specifications

Antigen Glucose Oxidase
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Glucose Oxidase.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Native purified Aspergillus niger Glucose Oxidase.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies recognize glucose oxidases from Aspergillus niger.
Target Species Bacteria, Fungi
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.