Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione, Mouse, Clone: D8, Abnova™

Catalog No. 89135265 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-265 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-265 Supplier Abnova Corporation Supplier No. MAB2829

Mouse monoclonal antibody raised against Glutathione.

Specifications

Antigen Glutathione
Applications Western Blot
Classification Monoclonal
Clone D8
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.01% sodium azide)
Host Species Mouse
Immunogen Glutathione-protein complexes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.