Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMPR, Mouse, Clone: 3G12, Abnova™

Catalog No. 89104554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-554 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-554 Supplier Abnova Corporation Supplier No. H00002766M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GMPR.

Guanosine monophosphate reductase (EC 1.7.1.7) catalyzes the irreversible NADPH-dependent reductive deamination of guanosine monophosphate (GMP) to inosine monophosphate (IMP). GMPR is able to convert guanosine nucleotides to the pivotal precursor of both guanine (G) and adenine (A) nucleotides. It plays an important role in maintaining the intracellular balance of A and G nucleotides.[supplied by OMIM

Sequence: MPRIDADLKLDFKDVLLRPKRSSLKSRAEVDLERTFTFRNSKQTYSGIPIIVANMDTVGTFEMAAVMSQHSMFTAIHKHYSLDDWKLFATNHPECLQNVAVSSGSGQNDLEKMTSILEAV

Specifications

Antigen GMPR
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GMPR.
Formulation ascites with no preservative
Gene GMPR
Gene Accession No. NM_006877
Gene Alias GMPR1
Gene Symbols GMPR
Host Species Mouse
Immunogen GMPR (NP_006868,1 a.a. ∽ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2766
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.