Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB5, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-385 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-385 Supplier Abnova Corporation Supplier No. H00010681A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GNB5.

Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq

Sequence: MCDQTFLVNVFGSCDKCFKQRALRPVFKKSQQLSYCSTCAEIMATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEAL

Specifications

Antigen GNB5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GNB5.
Formulation 50% glycerol
Gene GNB5
Gene Accession No. NM_016194
Gene Alias FLJ37457/FLJ43714/GB5
Gene Symbols GNB5
Host Species Mouse
Immunogen GNB5 (NP_057278, 1 a.a. to 90 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10681
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.