Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNGT2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104561 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-561 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-561 Supplier Abnova Corporation Supplier No. H00002793A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant GNGT2.

Phototransduction in rod and cone photoreceptors is regulated by groups of signaling proteins. The encoded protein is thought to play a crucial role in cone phototransduction. It belongs to the G protein gamma family and localized specifically in cones. There is evidence for use of multiple polyadenylation sites by this gene. [provided by RefSeq

Sequence: MAQDLSEKDLLKMEVEQLKKEVKNTRIPISKAGKEIKEYVEAQAGNDPFLKGIPEDKNPFKEKGGCLIS

Specifications

Antigen GNGT2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant GNGT2.
Formulation 50% glycerol
Gene GNGT2
Gene Accession No. BC008663
Gene Alias G-GAMMA-8/G-GAMMA-C/GNG8/GNG9/GNGT8
Gene Symbols GNGT2
Host Species Mouse
Immunogen GNGT2 (AAH08663, 1 a.a. to 69 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2793
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.