Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-GOT1, Clone: 1F5.2,

Catalog No. MABS508MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS508MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS508MI Supplier MilliporeSigma™ Supplier No. MABS508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects GOT1 clone: 1F5.2 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Aspartate aminotransferase, cytoplasmic (EC 2.6.2.1.; EC 2.6.2.3; UniProt P17174; also known as Aspartate aminotransferase 1, Aspartate transaminase 1, cAspAT, cCAT, Cysteine aminotransferase cytoplasmic, Cysteine transaminase cytoplasmic, Glutamate oxaloacetate transaminase 1, Growth-inhibiting protein 18, Transaminase A) is encoded by the GOT1 (also known as GIG18, ASTQTL1) gene (Gene ID 2805) in human. Aspartate aminotransferase is expressed in many types of cells and tissues and is conserved among prokaryotic and eukaryotic organisms. There exist two highly similar AST forms in mammals, the cytosolic cAST (encoded by the GOT1 gene) and the mitochondrial mAST (encoded by the GOT2 gene). Serum aspartate aminotransferase activity is largely that of cAST/GOT1, and elevated serum AST activity is associated with liver, heart, and muscle injury. AST catalyzes the reversible conversion between aspartate/alpha-ketoglutarate and oxaloacetate/glutamate, increasing oxaloacetate concentration therefore shifts the equilibrium of AST-catalzed reaction toward aspartate/alpha-ketoglutarate and away from glutatmate. Lowering blood glutamate concentration via oxaloacetate i.v. injection prompts an efflux of excess extracellular brain glutamate into the blood, which is reported to allevate the infarct size and edema in a rat model of ischemia induced by transient occlusion of the middle cerebral artery occlusion (MCAO).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F5.2
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P17174
Gene Symbols GOT1
Host Species Mouse
Immunogen GST-tagged Recombinant protein corresponding to the N-terminal region of human GOT1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002070
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.