Learn More
GOT2, Mouse, Clone: 14, Abnova™
Mouse monoclonal antibody raised against partial recombinant GOT2.
Supplier: Abnova Corporation MAB9901
Description
Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology. [provided by RefSeq
Specifications
GOT2 | |
Monoclonal | |
Unconjugated | |
Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
GOT2 | |
FLJ40994/KAT4/KATIV/mitAAT | |
Mouse | |
Affinity purification | |
RUO | |
2806 | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
14 | |
Mouse monoclonal antibody raised against partial recombinant GOT2. | |
In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
NM_002080.2 | |
GOT2 | |
Recombinant protein corresponding to amino acids 225-430 of human GOT2. | |
100 μg | |
Primary | |
It can expression in HepG2 whole cell lysate. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.