Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP1BA, Mouse, Clone: SZ2, Abnova™

Catalog No. 89111714 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-714 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-714 Supplier Abnova Corporation Supplier No. MAB4144
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native GP1BA.

Glycoprotein Ib (GP Ib) is a platelet surface membrane glycoprotein composed of a heterodimer, an alpha chain and a beta chain, that are linked by disulfide bonds. The Gp Ib functions as a receptor for von Willebrand factor (VWF). The complete receptor complex includes noncovalent association of the alpha and beta subunits with platelet glycoprotein IX and platelet glycoprotein V. The binding of the GP Ib-IX-V complex to VWF facilitates initial platelet adhesion to vascular subendothelium after vascular injury, and also initiates signaling events within the platelet that lead to enhanced platelet activation, thrombosis, and hemostasis. This gene encodes the alpha subunit. Several polymorphisms and mutations have been described in this gene, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease. [provided by RefSeq

Specifications

Antigen GP1BA
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone SZ2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native GP1BA.
Dilution Flow Cytometry (2ug/5x106 platelets/test) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (1mg/mL BSA)
Gene GP1BA
Gene Alias BSS/CD42B/CD42b-alpha/GP1B/MGC34595
Gene Symbols GP1BA
Host Species Mouse
Immunogen Native purified GP1BA from human washed platelets.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2811
Test Specificity The molecular weight of the recognized antigen is 160KDa. Reacts with glycoprotein Ib on megakaryocytes and platelets. Inhibits the ristocetin-dependent binding of Von Willebrand factor to platelets and ristocetin induced platelet agglutination. Inhibits platelet aggregation induced by Type I collagen and platelet activating factor (PAF). Immunoprecipitates the components of glycoprotein Ib complex.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.