Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPC3, Mouse, Clone: 1G12, Abnova™

Catalog No. 89108963 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-963 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-108-963 Supplier Abnova Corporation Supplier No. MAB10762
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against GPC3.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The protein encoded by this gene can bind to and inhibit the dipeptidyl peptidase activity of CD26, and it can induce apoptosis in certain cell types. Deletion mutations in this gene are associated with Simpson-Golabi-Behmel syndrome, also known as Simpson dysmorphia syndrome. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen GPC3
Applications Immunohistochemistry
Classification Monoclonal
Clone 1G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against GPC3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein base, 0.09% sodium azide)
Gene GPC3
Gene Alias DGSX/OCI-5/SDYS/SGB/SGBS/SGBS1
Gene Symbols GPC3
Host Species Mouse
Immunogen GPC3.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2719
Content And Storage Store at 4°C is stable for 3 years.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.