Learn More
GRHL2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GRHL2.
Supplier: Abnova Corporation H00079977A01
Description
The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of nonsyndromic sensorineural deafness autosomal dominant type 28 (DFNA28)
Sequence: ENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSSpecifications
GRHL2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant GRHL2. | |
GRHL2 | |
BOM/DFNA28/FLJ11172/FLJ13782/MGC149294/MGC149295/TFCP2L3 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_024915 | |
GRHL2 | |
GRHL2 (NP_079191, 111 a.a. to 210 a.a) partial recombinant protein with GST tag. | |
RUO | |
79977 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only