Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gria1, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89113375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-375 Supplier Abnova Corporation Supplier No. MAB8176
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against synthetic peptide of Gria1.

Specifications

Antigen Gria1
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Gria1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 0.1mg/mL BSA)
Gene Gria1
Gene Alias gluR-A
Gene Symbols Gria1
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of rat Gria1.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50592
Test Specificity This antibody has been directly tested for reactivity in Western blots with mouse and rat tissue. specific to the ∼105k GluR1 protein.
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.