Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIK1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104583 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-583 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-583 Supplier Abnova Corporation Supplier No. H00002897A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GRIK1.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. The subunit encoded by this gene is subject to RNA editing (CAG->CGG; Q->R) within the second transmembrane domain, which is thought to alter the properties of ion flow. Alternative splicing, resulting in transcript variants encoding different isoforms, has been noted for this gene. [provided by RefSeq

Sequence: VAIASHRASQLTVSSLQCHRHKPWRLGPRFMNLIKEARWDGLTGHITFNKTNGLRKDFDLDIISLKEEGTEKAAGEVSKHLYKVWKKIGIWNSNSGLNMTDSNKDKSSNI

Specifications

Antigen GRIK1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GRIK1.
Formulation 50% glycerol
Gene GRIK1
Gene Accession No. NM_000830
Gene Alias EAA3/EEA3/GLR5/GLUR5
Gene Symbols GRIK1
Host Species Mouse
Immunogen GRIK1 (NP_000821, 331 a.a. to 440 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2897
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.