Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grin1, Mouse, PerCP, Clone: S308-48, Abnova™

Catalog No. 89109859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-859 Supplier Abnova Corporation Supplier No. MAB11725
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial synthetic protein of Grin1.

Specifications

Antigen Grin1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S308-48
Conjugate PerCP
Description Mouse monoclonal antibody raised against partial synthetic protein of Grin1.
Dilution Immunohistochemistry (0.1-1 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS pH 7.4 (50% glycerol and 0.09% sodium azide)
Gene Grin1
Gene Alias NMDAR1/NR1
Gene Symbols Grin1
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to amino acids 42-361 at extracellular N-terminus of rat Grin1.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24408
Target Species Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.