Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIN3A/GRIN3B, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89136452 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-452 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-452 Supplier Abnova Corporation Supplier No. MAB8285
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant GRIN3A/GRIN3B.

Specifications

Antigen GRIN3A/GRIN3B
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant GRIN3A/GRIN3B.
Dilution Western Blot (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 780-914 of GRIN3A/GRIN3B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes NMDAR subunits GRIN3A and GRIN3B. It does not cross-react with subunits GRIN1 or GRIN2A.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.