Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK1, Mouse, Clone: 4E9, Abnova™

Catalog No. 89105334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-334 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-334 Supplier Abnova Corporation Supplier No. H00006011M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GRK1.

This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates rhodopsin and initiates its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). [provided by RefSeq

Sequence: RDSLSLEFESVCLEQPIGKKLFQQFLQSAEKHLPALELWKDIEDYDTADNDLQPQKAQTILAQYLDPQAKLFCSFLDEGIVAKFKEGPVEIQDGLFQPLL

Specifications

Antigen GRK1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GRK1.
Formulation ascites with no preservative
Gene GRK1
Gene Accession No. NM_002929
Gene Alias GPRK1/RHOK/RK
Gene Symbols GRK1
Host Species Mouse
Immunogen GRK1 (NP_002920.1,51 a.a. ∽ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6011
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.