Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK3A, Mouse, Clone: MS-1G7, Abnova™

Catalog No. 89108311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-311 Supplier Abnova Corporation Supplier No. MAB0989

Mouse monoclonal antibody raised against full-length recombinant GSK3A.

Glycogen synthase kinase 3-alpha (GSK3A; EC {2.7.1.37}) is a multifunctional protein serine kinase, homologous to Drosophila 'shaggy' (zeste-white3) and implicated in the control of several regulatory proteins including glycogen synthase (see GYS1, {138570}) and transcription factors (e.g., JUN, {165160}). It also plays a role in the WNT ({164820}) and PI3K (see PIK3CG, {601232}) signaling pathways (see review by {1:Ali et al. (2001)}).[supplied by OMIM

Specifications

Antigen GSK3A
Applications Western Blot
Classification Monoclonal
Clone MS-1G7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant GSK3A.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES/KOH, 150mM NaCl, pH 7.5 (50% glycerol)
Gene GSK3A
Gene Accession No. NM_019884
Gene Alias DKFZp686D0638
Gene Symbols GSK3A
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human GSK3A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2931
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.