Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK3A, Mouse, HRP, Clone: SBGSK3a1, Abnova™

Catalog No. 89136094 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-094 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-094 Supplier Abnova Corporation Supplier No. MAB6086
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant GSK3A.

Specifications

Antigen GSK3A
Applications Western Blot
Classification Monoclonal
Clone SBGSK3a1
Concentration Lot specific
Conjugate HRP
Description Mouse monoclonal antibody raised against recombinant GSK3A.
Dilution Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol)
Host Species Mouse
Immunogen Recombinant protein corresponding to GSK3A.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specificity human, mouse and rat GSK-3alpha
Target Species Human, Mouse, Rat
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.