Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gsk3a, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89109307 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-307 Supplier Abnova Corporation Supplier No. MAB11114

Mouse monoclonal antibody raised against recombinant protein of mouse Gsk3a.

Specifications

Antigen Gsk3a
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (0.01% sodium azide)
Host Species Mouse
Immunogen Recombinant Gsk3a from Xenopus laevis and mouse myeloma.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 606496/398338
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.