Learn More
GSK3B, Mouse, Clone: M131, Abnova™
Mouse monoclonal antibody raised against partial recombinant GSK3B.
Supplier: Abnova Corporation MAB1372
Description
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Specifications
GSK3B | |
Monoclonal | |
Unconjugated | |
ELISA (1:4000) Immunocytochemistry (1:1000) Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
GSK3B | |
Mouse | |
100 μL | |
Primary | |
This sequence is highly conserved in rat and mouse GSK-3b, but has low homology to GSK-3a. | |
Liquid |
ELISA, Western Blot | |
M131 | |
Mouse monoclonal antibody raised against partial recombinant GSK3B. | |
In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
GSK3B | |
Recombinant protein corresponding to N-terminus of human GSK3B. | |
RUO | |
2932 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.