Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST, Mouse, Clone: 1E5, Abnova™

Catalog No. 89134854 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-854 Supplier Abnova Corporation Supplier No. MAB1132
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant GST.

Specifications

Antigen GST
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant GST.
Dilution Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to full length GST .
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.