Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST, Mouse, Clone: 6G9B9, Abnova™

Catalog No. 89136334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-334 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-334 Supplier Abnova Corporation Supplier No. MAB7330

Mouse monoclonal antibody raised against recombinant GST.

Specifications

Antigen GST
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6G9B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant GST.
Dilution Western Blot (1:1000) Immunocytochemistry (1:200) Immunofluorescence (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.03% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to GST.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity GST (6G9B9).
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.